
Sulfadimethoxine

Synonym(s):
4-Amino-N-(2,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide, Sulfadimethoxine
Empirical Formula (Hill Notation):
C12H14N4O4S
CAS Number:
Molecular Weight:
310.33
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing