
(S)-Sitagliptin Phosphate

Synonym(s):
(S)-Sitagliptin Phosphate
Empirical Formula (Hill Notation):
C16H18F6N5O5P
CAS Number:
Molecular Weight:
505.31
Products
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing