
(S)-N-Fmoc-piperidine-2-carboxylic acid

Synonym(s):
Fmoc-Pip-OH, (S)-1-Fmoc-piperidine-2-carboxylic acid, N-Fmoc-L-pipecolinic acid
Empirical Formula (Hill Notation):
C21H21NO4
CAS Number:
Molecular Weight:
351.40
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing