
Sakakin

Synonym(s):
1-O-β-D-Glucosylorcinol
Empirical Formula (Hill Notation):
C13H18O7
CAS Number:
Molecular Weight:
286.28
Products
Product Category
Brand
Formula Weight
Physical Form
Research Area
Application
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing