
Rosiglitazone

Synonym(s):
Rosiglitazone, 5-[[4-[2-(Methyl-2-pyridinylamino)ethoxy]phenyl]methyl]-2,4-thiazolidinedione
Linear Formula:
C18H19N3O3S
CAS Number:
Molecular Weight:
357.43
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Agency
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing