
Rolipram

Synonym(s):
4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-2-pyrrolidinone, ZK 62711, Rolipram
Empirical Formula (Hill Notation):
C16H21NO3
CAS Number:
Molecular Weight:
275.34
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2