![Repaglinide Related Compound B certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/178/932/2390cc5f-bf6d-4752-a453-62f209b13d23/640/2390cc5f-bf6d-4752-a453-62f209b13d23.jpg)
Repaglinide Related Compound B
![Repaglinide Related Compound B certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/178/932/2390cc5f-bf6d-4752-a453-62f209b13d23/640/2390cc5f-bf6d-4752-a453-62f209b13d23.jpg)
Synonym(s):
[3-Ethoxy-4-(ethoxycarbonyl)phenyl]acetic acid
Empirical Formula (Hill Notation):
C13H16O5
CAS Number:
Molecular Weight:
252.26
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2