
Rapamycin

Synonym(s):
Rapamycin, 23,27-Epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine, AY 22989, Sirolimus
Empirical Formula (Hill Notation):
C51H79NO13
CAS Number:
Molecular Weight:
914.17
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Activity Spectrum
Format
Research Area
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3