
Ranitidine Related Compound B

Synonym(s):
N,N′-Bis{2-{{{5-[(dimethylamino)methyl]furan-2-yl}methyl}sulfanyl}ethyl}-2-nitroethene-1,1-diamine, N,N′-Bis{2-{{{5-[(dimethylamino)methyl]furan-2-yl}methyl}thio}ethyl}-2-nitro-1,1-ethylenediamine
Empirical Formula (Hill Notation):
C22H35N5O4S2
CAS Number:
Molecular Weight:
497.67
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2