Skip to Content
Merck
rac-BINAP 97%

rac-BINAP

rac-BINAP 97%
Synonym(s):
(±)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene, (±)-BINAP, [1,1′-Binaphthalene]-2,2′-diylbis[diphenylphosphine]
Linear Formula:
[(C6H5)2PC10H6-]2
CAS Number:
Molecular Weight:
622.67

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing