
Quizalofop-P-ethyl

Synonym(s):
DPX-Y6202, Ethyl (R)-2-[4-(6-chloro-2-quinoxalyloxy)phenoxy]propionate, Quinofop-ethyl
Empirical Formula (Hill Notation):
C19H17ClN2O4
CAS Number:
Molecular Weight:
372.80
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2