
Propylphosphonic anhydride solution

Synonym(s):
1-Propanephosphonic anhydride solution, 2,4,6-Tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide solution, PPACA, T3P®
Empirical Formula (Hill Notation):
C9H21O6P3
CAS Number:
Molecular Weight:
318.18
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing