
Propionylpromazine hydrochloride

Synonym(s):
Propiopromazine hydrochloride
Empirical Formula (Hill Notation):
C20H24N2OS · HCl
CAS Number:
Molecular Weight:
376.94
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing