Skip to Content
Merck
Potassium p-toluenethiosulfonate ≥97.0% (S)

Potassium p-toluenethiosulfonate

Potassium p-toluenethiosulfonate ≥97.0% (S)
Synonym(s):
p-Toluenethiosulfonic acid potassium salt, Potassium 4-methylbenzenethiosulfonate, Potassium thiotosylate
Linear Formula:
CH3C6H4SO2SK
CAS Number:
Molecular Weight:
226.36
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing