
Potassium persulfate

Synonym(s):
Potassium peroxodisulfate
Linear Formula:
K2S2O8
CAS Number:
Molecular Weight:
270.32
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Special Grade
Showing 1-3 of 3