
Potassium dihydrogen phosphate

Synonym(s):
Potassium phosphate monobasic, Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate
Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-2 of 2