
Oroxylin A

Synonym(s):
5,7-Dihydroxy-6-methoxy-2-phenyl-4H-1-benzopyran-4-one, 5,7-Dihydroxy-6-methoxyflavone;, 6-Methoxybaicalein;
Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing