Skip to Content
BA
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
petri dishes
Products
Product Category
dishes (3)
Manufacturer
Binding Type
Capacity (mL)
Diameter (mm)
Height (mm)
Surface Area cm²
Volume (ml)
Color
Package Size
Sterility
Sterilization Method
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
Z717223
Nunc
®
petri dishes,
diam. × H 90 mm × 15 mm, surface area 58 cm
2
, vented
Expand
View Pricing
Z717231
Nunc
®
petri dishes,
diam. × H 140 mm × 20 mm, surface area 145 cm
2
, vented
Expand
View Pricing
Z740356
Nunc
®
petri dishes,
H 150 mm × 20 mm, surface area × working volume 145 cm
2
× 35 mL, lid
Expand
View Pricing