Skip to Content
Merck
Norethindrone acetate United States Pharmacopeia (USP) Reference Standard

Norethindrone acetate

Norethindrone acetate United States Pharmacopeia (USP) Reference Standard
Synonym(s):
19-Norethindrone acetate, 17α-Ethynyl-17β-hydroxy-19-nor-4-androsten-3-one 17-acetate, 17α-Ethynyl-19-nortestosterone 17β-acetate, 17-Hydroxy-19-nor-17α-4-pregnen-20-yn-3-one acetate, 19-Norethisterone 17-acetate, 19-Norethisterone acetate
Empirical Formula (Hill Notation):
C22H28O3
CAS Number:
Molecular Weight:
340.46

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade