
Nitrofural

Synonym(s):
5-Nitro-2-furaldehyde semicarbazone, 5-Nitro-2-furfural semicarbazone, Furacin, NFZ, Nitrofurazone
Empirical Formula (Hill Notation):
C6H6N4O4
CAS Number:
Molecular Weight:
198.14
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing