
Neurotensin

Empirical Formula (Hill Notation):
C78H121N21O20
CAS Number:
Molecular Weight:
1672.92
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing