
Nefazodone Related Compound A

Synonym(s):
1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine monohydrochloride
Empirical Formula (Hill Notation):
C13H18Cl2N2 · HCl
CAS Number:
Molecular Weight:
309.66
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing