
Mupirocin calcium

Synonym(s):
Calcium mupirocin dihydrate
Empirical Formula (Hill Notation):
C52H86CaO18 · 2H2O
CAS Number:
Molecular Weight:
1075.34
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2