
Modafinil CIV

Synonym(s):
Modafinil, 2-[(Diphenylmethyl)sulfinyl]acetamide
Empirical Formula (Hill Notation):
C15H15NO2S
CAS Number:
Molecular Weight:
273.35
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing