
Milrinone Related Compound A

Synonym(s):
1,6-Dihydro-2-methyl-6-oxo(3,4′-bipyridine)-5-carboxamide, Milrinone amide
Empirical Formula (Hill Notation):
C12H11N3O2
CAS Number:
Molecular Weight:
229.23
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2