Skip to Content
Merck
Metolazone European Pharmacopoeia (EP) Reference Standard

Metolazone

Metolazone European Pharmacopoeia (EP) Reference Standard
Synonym(s):
7-Chloro-1,2,3,4-tetrahydro-2-methyl-3-(2-methylphenyl)-4-oxo-6-quinazolinesulfonamide
Empirical Formula (Hill Notation):
C16H16ClN3O3S
CAS Number:
Molecular Weight:
365.83

Products

Product Category
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade