Skip to Content
Merck
Methoxyamine hydrochloride derivatization grade (GC derivatization), LiChropur™, 97.5-102.5% (AT)

Methoxyamine hydrochloride

Methoxyamine hydrochloride derivatization grade (GC derivatization), LiChropur™, 97.5-102.5% (AT)
Synonym(s):
O-Methylhydroxylamine hydrochloride, Methoxylamine hydrochloride
Linear Formula:
CH3ONH2 · HCl
CAS Number:
Molecular Weight:
83.52

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reaction Type
Reagent Type
Technique