
Metam-sodium hydrate

Synonym(s):
Sodium methyldithiocarbamate
Empirical Formula (Hill Notation):
C2H4NNaS2 · xH2O
Molecular Weight:
129.18 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing