
Metalaxyl-M

Synonym(s):
N-(2,6-Dimethylphenyl)-N-(methoxyacetyl)-D-alanine methyl ester, Mefenoxam
Empirical Formula (Hill Notation):
C15H21NO4
CAS Number:
Molecular Weight:
279.33
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2