
Mercaptopurine impurity D

Synonym(s):
6,6′-Dithiodi-9H-purine
Empirical Formula (Hill Notation):
C10H6N8S2
CAS Number:
Molecular Weight:
302.34
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing