Skip to Content
Merck
Magnesium methyl carbonate solution 2.0 M in DMF

Magnesium methyl carbonate solution

Magnesium methyl carbonate solution 2.0 M in DMF
Synonym(s):
MMC, Methoxymagnesium methyl carbonate, Methyl methoxymagnesium carbonate, Stile’s reagent
Linear Formula:
CH3OMgOCO2CH3
CAS Number:
Molecular Weight:
130.38

Products

Product Category
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing