
Magnesium bis(trifluoromethanesulfonimide)

Empirical Formula (Hill Notation):
C4F12MgN2O8S4
CAS Number:
Molecular Weight:
584.60
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing