
Luperox® 531M80, 1,1-Bis(tert-amylperoxy)cyclohexane solution

Synonym(s):
1,1-Bis(tert-amylperoxy)cyclohexane solution
Linear Formula:
C6H10[OOC(CH3)2C2H5]2
CAS Number:
Molecular Weight:
288.42
Products
Product Category
Brand
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing