
L-Methionine-(methyl-d3)

Linear Formula:
CD3SCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
152.23
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing