
Linoleyl alcohol

Synonym(s):
cis,cis-9,12-Octadecadien-1-ol, Linoleyl alcohol
Empirical Formula (Hill Notation):
C18H34O
CAS Number:
Molecular Weight:
266.46
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2