
Linezolid Related Compound D

Synonym(s):
(5R)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-5-[[(methylsulfonyl)oxy]methyl]-2-oxazolidinone, (R)-[3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl]methyl methanesulfonate
Empirical Formula (Hill Notation):
C15H19FN2O6S
CAS Number:
Molecular Weight:
374.38
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2