
L-Arginine-(guanidineimino-15N2) hydrochloride

Synonym(s):
(S)-2-Amino-5-guanidinopentanoic acid-(guanidineimino-15N2), 15N Labeled L-arginine hydrochloride
Linear Formula:
H215NC(15NH)NH(CH2)3C(NH2)CO2H · HCl
CAS Number:
Molecular Weight:
212.65
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2