
L-Arginine-1-13C dihydrate

Empirical Formula (Hill Notation):
13CC5H14N4O2 · 2H2O
Molecular Weight:
211.22
Products
Product Category
Formula Weight
Physical Form
Purity
Isotope
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing