
Ketoprofen Related Compound C

Synonym(s):
3-[(1RS)-1-Carboxyethyl]benzoic acid, 2-(3-Carboxyphenyl)propionic acid, 3-Carboxy-α-methylbenzeneacetic acid
Empirical Formula (Hill Notation):
C10H10O4
CAS Number:
Molecular Weight:
194.18
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2