
Isopentenyl pyrophosphate trilithium salt

Synonym(s):
3-Methyl-3-butenyl pyrophosphate trilithium salt, IPP
Empirical Formula (Hill Notation):
C5H9Li3O7P2
CAS Number:
Molecular Weight:
263.89
Products
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing