
Iron(II) chloride tetrahydrate

Synonym(s):
Ferrous chloride tetrahydrate, Iron(II) chloride tetrahydrate
Linear Formula:
FeCl2 · 4H2O
CAS Number:
Molecular Weight:
198.81
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing