
Irgarol®

Synonym(s):
2-(tert-Butylamino)-4-(cyclopropylamino)-6-(methylthio)-s-triazine
Empirical Formula (Hill Notation):
C11H19N5S
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing