
Insulin Chain B Oxidized from bovine pancreas

Empirical Formula (Hill Notation):
C157H232N40O47S2
CAS Number:
Molecular Weight:
3495.89
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Package Size
Sterility
Technique
Showing 1-2 of 2