
Heptamethyl trisiloxane

Synonym(s):
1,1,1,3,5,5,5-Heptamethyltrisiloxane
Linear Formula:
[(CH3)3SiO]2SiHCH3
CAS Number:
Molecular Weight:
222.50
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing