
Heparin sodium porcine mucosa

Synonym(s):
Heparin sodium salt from porcine intestinal mucosa
CAS Number:
Products
Product Category
Biological Source
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing