
Hecogenin

Synonym(s):
12-Oxotigogenin, 3β-Hydroxy-5α-spirostan-12-one, 5α-Spirostan-3β-ol-12-one, Gekogenin, Hocogenin
Empirical Formula (Hill Notation):
C27H42O4
CAS Number:
Molecular Weight:
430.62
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing