
Flutamide Related Compound A

Synonym(s):
4-Nitro-3-(trifluoromethyl)aniline, 4-Nitro-α,α,α-trifluoro-m-toluidine, 5-Amino-2-nitrobenzotrifluoride
Linear Formula:
O2NC6H3(CF3)NH2
CAS Number:
Molecular Weight:
206.12
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing