Skip to Content
Merck
Flufenoxuron PESTANAL®, analytical standard

Flufenoxuron

Flufenoxuron PESTANAL®, analytical standard
Synonym(s):
N-{4-[2-Chloro-4-(trifluoromethyl)phenoxy]-2-fluorophenylaminocarbonyl}-2,6-difluoro-benzamide
Empirical Formula (Hill Notation):
C21H11ClF6N2O3
CAS Number:
Molecular Weight:
488.77
EC Number:

Products

Product Category
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique