
Faradiol

Synonym(s):
(3β,16β,18α,19α)-Urs-20-ene-3,16-diol, 16β-Hydroxypseudotaraxasterol
Empirical Formula (Hill Notation):
C30H50O2
CAS Number:
Molecular Weight:
442.72
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing