Skip to Content
Merck
Ethylenediaminetetraacetic acid ferric sodium salt suitable for plant cell culture, BioReagent, crystalline

Ethylenediaminetetraacetic acid ferric sodium salt

Ethylenediaminetetraacetic acid ferric sodium salt suitable for plant cell culture, BioReagent, crystalline
Synonym(s):
Ethylenediaminetetraacetic acid iron(III) sodium salt, EDTA iron(III) sodium salt, Edathamil, Komplexon II
Linear Formula:
C10H12N2NaFeO8
CAS Number:
Molecular Weight:
367.05
EC Number:

Products

Product Category
Formula Weight
Physical Form
Product Line
Technique